Order Entry
Ireland
ContactUsLinkComponent
Mouse PPP2R1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse PPP2R1A in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA270920-96
Supplier:  ANTIBODIES.COM
New Product
Mouse PPP2R1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse PPP2R1A in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA270920-96
Supplier:  ANTIBODIES.COM