Order Entry
Ireland
ContactUsLinkComponent
Mouse Lipocalin-2/NGAL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse Lipocalin-2/NGAL in serum, plasma, tissue homogenates, urine, cell lysates, cell culture supernat 1 * 96 Tests
Catalog # ANTIA2366-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Lipocalin-2/NGAL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse Lipocalin-2/NGAL in serum, plasma, tissue homogenates, urine, cell lysates, cell culture supernat 1 * 96 Tests
Catalog # ANTIA2366-96
Supplier:  ANTIBODIES.COM