Order Entry
Ireland
ContactUsLinkComponent
Human Complement C8a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Complement C8a in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270656-48
Supplier:  ANTIBODIES.COM
New Product
Human Complement C8a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Complement C8a in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270656-48
Supplier:  ANTIBODIES.COM