Canine Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-AP, recombinant canine Interferon gamma ELISA standard, and standard reconstitution buffe 1 * 576 Tests
Catalog # ANTIA270377-696
Supplier: ANTIBODIES.COM