Order Entry
Ireland
ContactUsLinkComponent
Rat ITLN1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat ITLN1 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2518-96
Supplier:  ANTIBODIES.COM
New Product
Rat ITLN1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat ITLN1 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2518-96
Supplier:  ANTIBODIES.COM