Order Entry
Ireland
ContactUsLinkComponent
Human Angiopoietin 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Angiopoietin 1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA1651-48
New Product
Human Angiopoietin 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Angiopoietin 1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA1651-48