Order Entry
Ireland
ContactUsLinkComponent
Human Surfactant Protein A/PSAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Surfactant Protein A/PSAP in serum, plasma, tissue homogenates, cell lysates, lung lavag 1 * 48 Tests
Catalog # ANTIA2009-48
Supplier:  ANTIBODIES.COM
New Product
Human Surfactant Protein A/PSAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Surfactant Protein A/PSAP in serum, plasma, tissue homogenates, cell lysates, lung lavag 1 * 48 Tests
Catalog # ANTIA2009-48
Supplier:  ANTIBODIES.COM