Order Entry
Ireland
ContactUsLinkComponent
Human Complement C4b ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Complement C4b in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2326-96
Supplier:  ANTIBODIES.COM
New Product
Human Complement C4b ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Complement C4b in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2326-96
Supplier:  ANTIBODIES.COM