Order Entry
Ireland
ContactUsLinkComponent
Human Anti-SARS-CoV-2 Spike RBD IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 Spike RBD IgG in serum, plasma, cell culture supernatant, cell or tissue lysa 1 * 96 Tests
Catalog # ANTIA326397-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-SARS-CoV-2 Spike RBD IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 Spike RBD IgG in serum, plasma, cell culture supernatant, cell or tissue lysa 1 * 96 Tests
Catalog # ANTIA326397-96
Supplier:  ANTIBODIES.COM