Order Entry
Ireland
ContactUsLinkComponent
Human Cytokeratin 17 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Cytokeratin 17 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA2857-48
Supplier:  ANTIBODIES.COM
New Product
Human Cytokeratin 17 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Cytokeratin 17 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA2857-48
Supplier:  ANTIBODIES.COM