Order Entry
Ireland
ContactUsLinkComponent
Human Tau (phospho T217) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Tau (phospho T217) in serum, plasma, cell culture supernatant, cell or tissue lysate, and other 1 * 96 Tests
Catalog # ANTIA326853-96
Supplier:  ANTIBODIES.COM
New Product
Human Tau (phospho T217) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Tau (phospho T217) in serum, plasma, cell culture supernatant, cell or tissue lysate, and other 1 * 96 Tests
Catalog # ANTIA326853-96
Supplier:  ANTIBODIES.COM