Order Entry
Ireland
ContactUsLinkComponent
Human Serum Indoxyl Sulfate ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantita tive determination of human Serum Indoxyl Sulfate in serum, plasma, cell culture supernatant, cell or tissue lysate, 1 * 96 Tests
Catalog # ANTIA326683-96
Supplier:  ANTIBODIES.COM
New Product
Human Serum Indoxyl Sulfate ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantita tive determination of human Serum Indoxyl Sulfate in serum, plasma, cell culture supernatant, cell or tissue lysate, 1 * 96 Tests
Catalog # ANTIA326683-96
Supplier:  ANTIBODIES.COM