Order Entry
Ireland
ContactUsLinkComponent
Hamster IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, native Syrian hamster IgG ELISA standard, and standard reconstitution buffer. This matched antib 1 * 576 Tests
Catalog # ANTIA325363-696
Supplier:  ANTIBODIES.COM
New Product
Hamster IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, native Syrian hamster IgG ELISA standard, and standard reconstitution buffer. This matched antib 1 * 576 Tests
Catalog # ANTIA325363-696
Supplier:  ANTIBODIES.COM