Order Entry
Ireland
ContactUsLinkComponent
Mouse EFEMP1/Fibulin-3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse EFEMP1/Fibulin-3 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liq 1 * 96 Tests
Catalog # ANTIA326560-96
Supplier:  ANTIBODIES.COM
New Product
Mouse EFEMP1/Fibulin-3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse EFEMP1/Fibulin-3 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liq 1 * 96 Tests
Catalog # ANTIA326560-96
Supplier:  ANTIBODIES.COM