Order Entry
Ireland
ContactUsLinkComponent
Human VEGFC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human VEGFC in serum, platelet-poor plasma, tissue homogenates, cell lysates, cell culture supernates or other biol 1 * 96 Tests
Catalog # ANTIA3084-96
Supplier:  ANTIBODIES.COM
New Product
Human VEGFC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human VEGFC in serum, platelet-poor plasma, tissue homogenates, cell lysates, cell culture supernates or other biol 1 * 96 Tests
Catalog # ANTIA3084-96
Supplier:  ANTIBODIES.COM