Order Entry
Ireland
ContactUsLinkComponent
Human BAFF-R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human BAFF-R in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3126-96
Supplier:  ANTIBODIES.COM
New Product
Human BAFF-R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human BAFF-R in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3126-96
Supplier:  ANTIBODIES.COM