Order Entry
Ireland
ContactUsLinkComponent
Human Anti-Myelin Basic Protein Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantit ative determination of human Anti-Myelin Basic Protein Antibody in serum, plasma, tissue homogenates, and other biolo 1 * 96 Tests
Catalog # ANTIA303123-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-Myelin Basic Protein Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantit ative determination of human Anti-Myelin Basic Protein Antibody in serum, plasma, tissue homogenates, and other biolo 1 * 96 Tests
Catalog # ANTIA303123-96
Supplier:  ANTIBODIES.COM