Order Entry
Ireland
ContactUsLinkComponent
Horse Enkephalin/ENK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of horse Enkephalin/ENK in serum, plasma, saliva or other biological fluids 1 * 96 Tests
Catalog # ANTIA326181-96
Supplier:  ANTIBODIES.COM
New Product
Horse Enkephalin/ENK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of horse Enkephalin/ENK in serum, plasma, saliva or other biological fluids 1 * 96 Tests
Catalog # ANTIA326181-96
Supplier:  ANTIBODIES.COM