Order Entry
Ireland
ContactUsLinkComponent
Hamster Prolactin/PRL ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative d etermination of hamster Prolactin/PRL in serum, plasma, cell culture supernatant, cell or tissue lysate, and other li 1 * 96 Tests
Catalog # ANTIA326843-96
Supplier:  ANTIBODIES.COM
New Product
Hamster Prolactin/PRL ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative d etermination of hamster Prolactin/PRL in serum, plasma, cell culture supernatant, cell or tissue lysate, and other li 1 * 96 Tests
Catalog # ANTIA326843-96
Supplier:  ANTIBODIES.COM