Order Entry
Ireland
ContactUsLinkComponent
Mouse UCN2/SRP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse UCN2/SRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA4945-48
Supplier:  ANTIBODIES.COM
New Product
Mouse UCN2/SRP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse UCN2/SRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA4945-48
Supplier:  ANTIBODIES.COM