Order Entry
Ireland
ContactUsLinkComponent
Mouse VLDL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse VLDL in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5140-96
Supplier:  ANTIBODIES.COM
New Product
Mouse VLDL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse VLDL in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5140-96
Supplier:  ANTIBODIES.COM