Order Entry
Ireland
ContactUsLinkComponent
Porcine Alanine Transaminase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of porcine Alanine Transaminase in serum, plasma, tissue homogenates, cell lysates, cell culture sup 1 * 96 Tests
Catalog # ANTIA3355-96
Supplier:  ANTIBODIES.COM
New Product
Porcine Alanine Transaminase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of porcine Alanine Transaminase in serum, plasma, tissue homogenates, cell lysates, cell culture sup 1 * 96 Tests
Catalog # ANTIA3355-96
Supplier:  ANTIBODIES.COM