Order Entry
Ireland
ContactUsLinkComponent
Mouse FGF6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse FGF6 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3398-48
Supplier:  ANTIBODIES.COM
New Product
Mouse FGF6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse FGF6 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3398-48
Supplier:  ANTIBODIES.COM