Order Entry
Ireland
ContactUsLinkComponent
Horse Procollagen II N-Terminal Propeptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vi tro quantitative determination of horse Procollagen II N-Terminal Propeptide in serum, plasma or other biological flu 1 * 96 Tests
Catalog # ANTIA4442-96
Supplier:  ANTIBODIES.COM
New Product
Horse Procollagen II N-Terminal Propeptide ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vi tro quantitative determination of horse Procollagen II N-Terminal Propeptide in serum, plasma or other biological flu 1 * 96 Tests
Catalog # ANTIA4442-96
Supplier:  ANTIBODIES.COM