Order Entry
Ireland
ContactUsLinkComponent
Mouse UQCRC2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse UQCRC2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4939-48
Supplier:  ANTIBODIES.COM
New Product
Mouse UQCRC2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse UQCRC2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4939-48
Supplier:  ANTIBODIES.COM