Order Entry
Ireland
ContactUsLinkComponent
Mouse Huntingtin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Huntingtin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA5652-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Huntingtin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Huntingtin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA5652-96
Supplier:  ANTIBODIES.COM