Order Entry
Ireland
ContactUsLinkComponent
Tetracycline Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Tetracycline in honey. 1 * 48 Tests
Catalog # ANTIA327258-48
Supplier:  ANTIBODIES.COM
New Product
Tetracycline Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Tetracycline in honey. 1 * 48 Tests
Catalog # ANTIA327258-48
Supplier:  ANTIBODIES.COM