Order Entry
Ireland
ContactUsLinkComponent
Rabbit S100 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of rabbit S100 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA3280-96
Supplier:  ANTIBODIES.COM
New Product
Rabbit S100 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of rabbit S100 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA3280-96
Supplier:  ANTIBODIES.COM