Order Entry
Ireland
ContactUsLinkComponent
Mouse Tau ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Tau in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4727-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Tau ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Tau in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4727-48
Supplier:  ANTIBODIES.COM