Human OAS1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human OAS1 in serum, plasma, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4969-48
Supplier: ANTIBODIES.COM