Order Entry
Ireland
ContactUsLinkComponent
Rat Motilin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Motilin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5027-48
Supplier:  ANTIBODIES.COM
New Product
Rat Motilin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Motilin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5027-48
Supplier:  ANTIBODIES.COM