Order Entry
Ireland
ContactUsLinkComponent
Human SERCA2 ATPase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human SERCA2 ATPase in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5208-96
Supplier:  ANTIBODIES.COM
New Product
Human SERCA2 ATPase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human SERCA2 ATPase in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5208-96
Supplier:  ANTIBODIES.COM