Order Entry
Ireland
ContactUsLinkComponent
Rat IGFBP5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat IGFBP5 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA4205-96
New Product
Rat IGFBP5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat IGFBP5 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA4205-96