Order Entry
Ireland
ContactUsLinkComponent
Human CD83 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human CD83 in serum, plasma, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA5039-96
New Product
Human CD83 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human CD83 in serum, plasma, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA5039-96