Order Entry
Ireland
ContactUsLinkComponent
Rat IL-23R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat IL-23R in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5166-48
Supplier:  ANTIBODIES.COM
New Product
Rat IL-23R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat IL-23R in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5166-48
Supplier:  ANTIBODIES.COM