Order Entry
Ireland
ContactUsLinkComponent
Human Chromogranin B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Chromogranin B in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5135-96
Supplier:  ANTIBODIES.COM
New Product
Human Chromogranin B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Chromogranin B in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5135-96
Supplier:  ANTIBODIES.COM