Order Entry
Ireland
ContactUsLinkComponent
Chicken BMP4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of chicken BMP4 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4505-96
Supplier:  ANTIBODIES.COM
New Product
Chicken BMP4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of chicken BMP4 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4505-96
Supplier:  ANTIBODIES.COM