Order Entry
Ireland
ContactUsLinkComponent
Human Periostin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Periostin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4855-48
Supplier:  ANTIBODIES.COM
New Product
Human Periostin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Periostin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4855-48
Supplier:  ANTIBODIES.COM