Order Entry
Ireland
ContactUsLinkComponent
Human IL-17A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-17A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 96 Tests
Catalog # ANTIA3581-96
Supplier:  ANTIBODIES.COM
New Product
Human IL-17A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-17A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 96 Tests
Catalog # ANTIA3581-96
Supplier:  ANTIBODIES.COM