Order Entry
Ireland
ContactUsLinkComponent
Human Biglycan ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Biglycan in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5494-96
Supplier:  ANTIBODIES.COM
New Product
Human Biglycan ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Biglycan in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5494-96
Supplier:  ANTIBODIES.COM