Monkey Fas Ligand ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of monkey Fas Ligand in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
ANTIA4541-48