Order Entry
Ireland
ContactUsLinkComponent
Rat Albumin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinati on of rat Albumin in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3753-96
Supplier:  ANTIBODIES.COM
New Product
Rat Albumin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinati on of rat Albumin in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3753-96
Supplier:  ANTIBODIES.COM