Order Entry
Ireland
ContactUsLinkComponent
Fumonisin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Fumonisin in feed, cereals sample. 1 * 50 Tests
Catalog # ANTIA327211-50
Supplier:  ANTIBODIES.COM
New Product
Fumonisin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Fumonisin in feed, cereals sample. 1 * 50 Tests
Catalog # ANTIA327211-50
Supplier:  ANTIBODIES.COM