Order Entry
Ireland
ContactUsLinkComponent
Mouse NMDAR2A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse NMDAR2A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4963-48
Supplier:  ANTIBODIES.COM
New Product
Mouse NMDAR2A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse NMDAR2A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4963-48
Supplier:  ANTIBODIES.COM