Order Entry
Ireland
ContactUsLinkComponent
Homovanillic Acid ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative deter mination of Homovanillic Acid in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4706-96
Supplier:  ANTIBODIES.COM
New Product
Homovanillic Acid ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative deter mination of Homovanillic Acid in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4706-96
Supplier:  ANTIBODIES.COM