Order Entry
Ireland
ContactUsLinkComponent
Mouse OCT4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse OCT4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3885-48
Supplier:  ANTIBODIES.COM
New Product
Mouse OCT4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse OCT4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3885-48
Supplier:  ANTIBODIES.COM