Order Entry
Ireland
ContactUsLinkComponent
Horse MMP7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f horse MMP7 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3937-96
Supplier:  ANTIBODIES.COM
New Product
Horse MMP7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f horse MMP7 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3937-96
Supplier:  ANTIBODIES.COM