Order Entry
Ireland
ContactUsLinkComponent
Human e-LOX-3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human e-LOX-3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5689-48
Supplier:  ANTIBODIES.COM
New Product
Human e-LOX-3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human e-LOX-3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5689-48
Supplier:  ANTIBODIES.COM