Order Entry
Ireland
ContactUsLinkComponent
Chicken IL-10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of chicken IL-10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
  ANTIA3877-48
New Product
Chicken IL-10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of chicken IL-10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
  ANTIA3877-48