Order Entry
Ireland
ContactUsLinkComponent
Mouse SERCA2 ATPase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse SERCA2 ATPase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5732-96
Supplier:  ANTIBODIES.COM
New Product
Mouse SERCA2 ATPase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse SERCA2 ATPase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5732-96
Supplier:  ANTIBODIES.COM